missing translation for 'onlineSavingsMsg'
Learn More

IKA™ myPlate Magnetic Stirrer
GREENER_CHOICE

IKA myPlate Magnetic Stirrer with personal interchangeable frame and easy-to-use design that allows you to set it up by yourself.

Brand:  IKA™ 0025005156

404.70 EUR valid until 2025-12-31
Use promo code "24299" to get your promotional price.


Product Code. 16110361

  • 426.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Device equipped with a decorative image template that can be exchanged easily through the convenient interchangeable frame
  • Allows you to convert the magnetic stirrer into a canvas of pictures
  • High chemical resistance
  • Electronically controlled motor
  • Digital speed indicator
Specifications

Specifications

100 to 2500 rpm
135 mm
100 to 240 V, 50/60 Hz, 4 W, 300 mA
Rotates to the left
0.76 kg
5.3 in.
30 x 152 x 164 mm
Speed adjustment push-button
5°C to 40°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.