missing translation for 'onlineSavingsMsg'
Learn More

Myricetin, 95%

Product Code. 10745991 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mg
Packaging:
Glass bottle
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 10745991

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 529-44-2 | C15H10O8 | 318.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 529-44-2
CAS Min % 94.0
CAS Max % 100.0
Color Brown-Green to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C15H10O8
MDL Number MFCD00006827
Quantity 25 mg
Beilstein 18, 257
Merck Index 15, 6417
Synonym myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
InChI Key IKMDFBPHZNJCSN-UHFFFAOYSA-N
SMILES OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C(O)=C2)=C1
IUPAC Name 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
Molecular Weight (g/mol) 318.24
PubChem CID 5281672
ChEBI CHEBI:18152
Formula Weight 318.23
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material Myricetin, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.