Learn More
VACUUBRAND™ MZ 2 NT Diaphragm Pump
GREENER_CHOICE
An excellent solution for continuous, oil-free evacuation and pumping of gases.
Brand:  VACUUBRAND™ 20732002
Product Code. 11929961
  • 1715.00 € / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • All parts in contact with pumped media are made of aluminum and selected plastics and allow a wide range of applications for non-corrosive gas
  • Two-stage construction provides the advantageous combination of high pumping speed and an ultimate vacuum down to 7 mbar
  • Highly flexible fabric-reinforced double diaphragm made of FKM is ideal for extended operating life
  • Contamination-free pumping and evacuation
  • High flow rate even near ultimate vacuum
  • Whisper quiet and very low vibration
  • Exceptionally long diaphragm life and maintenance-free drive system for low lifetime costs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7 mbar
MZ 2 NT
2.2 m³ at 50Hz; 2.4 m³ at 60Hz
243 x 239 x 198 mm
11 kg