missing translation for 'onlineSavingsMsg'
Learn More

n-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals™

Product Code. p-7083046 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g

Product Code. 11423811

Brand: MP Biomedicals 0215125201

Please to purchase this item. Need a web account? Register with us today!

Used as an ion pairing reagent for HPLC and as an anionic surfactant. n-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals has a low molecular weight that will not form micelles in solution.

Sodium hexanesulfonate is utilized in the analysis of small organic compounds, and pharmaceutical products and metabolites by such techniques as HPLC.

  • The anionic sulfonate counterion permits the separation and resolution of positively charged analytes
  • Because of its low molecular weight, it will not form micelles in solution and therefore has no CMC value
  • For ion-pair chromatography
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2–8°C
Quantity 1 g
Molecular Formula C6H13NaO3S
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
SMILES [Na+].CCCCCCS([O-])(=O)=O
IUPAC Name sodium hexane-1-sulfonate
Molecular Weight (g/mol) 188.22
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.