missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine dihydrochloride, 96%, Thermo Scientific Chemicals

Product Code. 11403387
Click to view available options
Quantity:
10 g
50 g

N-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.

Solubility
Soluble in hot water, alcohol, dilute hydrochloric acid and acetone.

Notes
Hygroscopic. Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, halogen acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.174
MDL Number MFCD00012556
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
PubChem CID 15106
ChEBI CHEBI:53452
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl

Specifications

Melting Point 191°C to 199°C
Quantity 50 g
Beilstein 3707471
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,6409
Solubility Information Soluble in hot water,alcohol,dilute hydrochloric acid and acetone.
Formula Weight 259.18
Percent Purity 96%
Chemical Name or Material N-(1-Naphthyl)ethylenediamine dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'