missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Decomposition Information | 200°C |
Color | Beige to White or Pink |
Infrared Spectrum | Authentic |
Assay Percent Range | 98+% |
Packaging | Glass Bottle |
Molecular Formula | C12H16Cl2N2 |
Linear Formula | C10H7NHCH2CH2NH2·2HCl |
Quantity | 25 g |
Show More |
Safety and Handling
Product Identifier
- N-(1-Naphthyl)ethylenediamine, dihydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'