missing translation for 'onlineSavingsMsg'
Learn More

N-1-Naphthylethylene Diamine Dihydrochloride, Extra Pure, SLR, Fisher Chemical™

Product Code. 10704191
Click to view available options
Quantity:
10 g
100 g

N-1-Naphthylethylene Diamine Dihydrochloride, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.174
MDL Number 12556
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
PubChem CID 15106
ChEBI CHEBI:53452
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl

Specifications

Melting Point 200°C
Color Beige
Packaging Glass Jar
Quantity 100 g
Formula Weight 259.17g/mol
Physical Form Solid
compliance-icons
Product Identifier
  • N-(1-Naphthyl)ethylenediamine, dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.