missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Difluorophenyl)thiourea, 97%, Thermo Scientific™

Product Code. 11428786
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175277-76-6
Molecular Formula C7H6F2N2S
Molecular Weight (g/mol) 188.196
MDL Number MFCD00041150
InChI Key DZZSKQFBAGZNSH-UHFFFAOYSA-N
Synonym 2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl-2-thiourea, n-2,4-difluorophenyl thiourea, thiourea,n-2,4-difluorophenyl, amino 2,4-difluorophenyl amino methane-1-thione, acmc-1bxvl, buttpark 12118-31, difluorophenylthioacetonitrile, 2,4-difluoro-phenyl-thiourea
PubChem CID 2734207
IUPAC Name (2,4-difluorophenyl)thiourea
SMILES C1=CC(=C(C=C1F)F)NC(=S)N

Specifications

Melting Point 165°C to 168°C
Quantity 1 g
UN Number UN2811
Formula Weight 188.2
Percent Purity 97%
Chemical Name or Material N-(2,4-Difluorophenyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'