missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dimethylphenyl)thiourea, 98%, Thermo Scientific™

Product Code. 11318675
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16738-20-8
Molecular Formula C9H12N2S
Molecular Weight (g/mol) 180.269
MDL Number MFCD00041164
InChI Key KAGLPYRXTCQWHU-UHFFFAOYSA-N
Synonym 1-2,4-dimethylphenyl-2-thiourea, 2,4-dimethylphenyl thiourea, 2,4-xylylthiourea, n-2,4-dimethylphenyl thiourea, 1-2,4-dimethylphenyl thiourea, 2,4-xylyl thiourea, 2-thio-1-2,4-xylyl urea, urea, 2-thio-1-2,4-xylyl, thiourea, 2,4-dimethylphenyl-9ci, thiourea,n-2,4-dimethylphenyl
PubChem CID 2798617
IUPAC Name (2,4-dimethylphenyl)thiourea
SMILES CC1=CC(=C(C=C1)NC(=S)N)C

Specifications

Melting Point 177°C to 180°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 2639862
Formula Weight 180.27
Percent Purity 98%
Chemical Name or Material N-(2,4-Dimethylphenyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'