missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminoethyl)piperidine, 98%, Thermo Scientific Chemicals

Product Code. 10636544
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27578-60-5
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD00006516
InChI Key CJNRGSHEMCMUOE-UHFFFAOYSA-N
Synonym n-2-aminoethyl piperidine, 1-2-aminoethyl piperidine, 2-piperidin-1-yl ethanamine, 2-piperidin-1-yl ethan-1-amine, 1-piperidineethanamine, 2-piperidinoethylamine, 2-1-piperidinyl ethanamine, 2-1-piperidinyl ethylamine, n-aminoethylpiperidine, 2-piperidino-1-ethanamine
PubChem CID 33944
IUPAC Name 2-piperidin-1-ylethanamine
SMILES C1CCN(CC1)CCN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 186°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4730 to 1.475
Quantity 25 mL
Beilstein 20, 67
Specific Gravity 0.89
Formula Weight 128.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-(2-Aminoethyl)piperidine
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'