missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Bromophenyl)thiourea, 98%, Thermo Scientific Chemicals

Product Code. 11342437
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5391-30-0
Molecular Formula C7H7BrN2S
Molecular Weight (g/mol) 231.111
MDL Number MFCD00041146
InChI Key QIGMVYSPXPXCPN-UHFFFAOYSA-N
Synonym 1-2-bromophenyl thiourea, 1-2-bromophenyl-2-thiourea, 2-bromophenyl thiourea, n-2-bromophenyl thiourea, thiourea, 2-bromophenyl, thiourea,n-2-bromophenyl, thiourea, bromophenyl, amino 2-bromophenyl amino methane-1-thione, o-bromophenylthiourea
PubChem CID 2735618
IUPAC Name (2-bromophenyl)thiourea
SMILES C1=CC=C(C(=C1)NC(=S)N)Br

Specifications

Melting Point 124°C to 126°C
Odor Odorless
Quantity 1 g
UN Number UN2811
Beilstein 2718427
Formula Weight 231.12
Percent Purity 98%
Chemical Name or Material N-(2-Bromophenyl)thiourea

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'