missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Chloro-3-pyridyl)maleimide

Product Code. p-7033433
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11430000

Brand: Alfa Aesar™ H50547.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 278610-39-2
Molecular Formula C9H5ClN2O2
Molecular Weight (g/mol) 208.60
MDL Number MFCD00111928
InChI Key JVJJPDZQLVCNFU-UHFFFAOYSA-N
Synonym n-2-chloro-3-pyridyl maleimide, 1-2-chloro-3-pyridyl-2,5-dihydro-1h-pyrrole-2,5-dione, 1-2-chloropyridin-3-yl pyrrole-2,5-dione, 1-2-chloropyridin-3-yl-1h-pyrrole-2,5-dione, 1-2-chloropyridin-3-yl-2,5-dihydro-1h-pyrrole-2,5-dione, n-2-chloropyridin-3-yl maleimide, 1-2-chloro-3-pyridyl azoline-2,5-dione, n-2-chloropyridine-3-yl maleimide
PubChem CID 2781068
IUPAC Name 1-(2-chloropyridin-3-yl)pyrrole-2,5-dione
SMILES ClC1=C(C=CC=N1)N1C(=O)C=CC1=O

Specifications

Quantity 1 g
Formula Weight 208.61
Chemical Name or Material N-(2-Chloro-3-pyridyl)maleimide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.