missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Pyridyl)thiourea, 98%, Thermo Scientific Chemicals

Product Code. 11307796
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-(2-Pyridyl)thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Pyridyl)thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14294-11-2
Molecular Formula C6H7N3S
Molecular Weight (g/mol) 153.20
MDL Number MFCD00041227
InChI Key SLUHLANJIVXTRQ-UHFFFAOYSA-N
Synonym 1-pyridin-2-yl thiourea, 2-pyridylthiourea, 1-pyridin-2-ylthiourea, 1-2-pyridyl-2-thiourea, n-2-pyridinyl-thiourea, pyridin-2-yl thiourea, pyridin-2-yl-thiourea, n-pyridin-2-ylthiourea, n-2-pyridinyl thiourea, 2-pyridyl thiourea
PubChem CID 1490491
IUPAC Name pyridin-2-ylthiourea
SMILES NC(=S)NC1=CC=CC=N1

Specifications

Melting Point 146°C to 149°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 117849
Formula Weight 153.21
Percent Purity 98%
Chemical Name or Material N-(2-Pyridyl)thiourea

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'