missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ N-2 Supplement (100X)

Product Code. 15410294
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mL
50 mL
missing translation for 'unitSize'
50mL
5mL
This item is not returnable. View return policy

Product Code. 15410294

missing translation for 'mfr': Gibco™ 17502001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-2 Supplement is a versatile serum-free cell culture supplement that supports the growth and viability of various cell types.

Gibco N-2 Supplement is a chemically defined, serum-free supplement based on Bottenstein's N-1 formulation, containing optimized concentrations of insulin, iron-rich holo-transferrin, progesterone, putrescine, and selenite. N-2 Supplement is recommended for growth and expression of neuroblastomas and neural cells in primary cultures from both the peripheral nervous system (PNS) and the central nervous system (CNS).

N-2 Supplement has been demonstrated in wide-ranging applications, such as the culture of neural stem cells, the culture of the SH-SY5Y cell line (subline of the neuroblastoma cell line SK-N-SH), and culture of the HT22 mouse hippocampal neuronal cell line. Additionally, N-2 Supplement adds support to cell culture media used to differentiate pluripotent stem cells (PSCs) to a variety of cell types, including neural and pancreatic-like cell types.

N-2 Supplement may be used with Neurobasal media supplemented with growth factors such as bFGF and EGF. When combined with Dulbecco's Modified Eagle Medium (D-MEM) and fetal bovine serum (FBS), N-2 Supplement also provides suitable supplementation for the culture of human and rodent astrocytes.

  • Each lot is cGMP manufactured
  • Each lot is tested for sterility, endotoxin, and performance
  • Contains insulin, iron-rich holo-transferrin, progesterone, putrescine, and selenite
  • Can be used as a substitute for Bottenstein's N-1 formulation
  • Provided as a 100X concentrate

Choose Gibco N-2 Supplement for a reliable, high-quality product trusted by researchers worldwide.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -20°C) and protect from light.

Cell Type Neural Cells
Classification Serum-free
Concentration 100 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
pH 6 to 8
Quantity 50 mL
Shipping Condition Dry Ice
Source Serum-Free
Tested For Endotoxin, Performance, Sterility
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.