missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ N-2 Supplement (100X)

Product Code. p-7076112
Click to view available options
Quantity:
5 mL
50 mL
Unit Size:
50mL
5mL
This item is not returnable. View return policy

Product Code. 11520536

Brand: Gibco™ 17502048

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-2 Supplement is a versatile serum-free cell culture supplement that supports the growth and viability of various cell types.

Gibco N-2 Supplement is a chemically defined, serum-free supplement based on Bottenstein's N-1 formulation, containing optimized concentrations of insulin, iron-rich holo-transferrin, progesterone, putrescine, and selenite. N-2 Supplement is recommended for growth and expression of neuroblastomas and neural cells in primary cultures from both the peripheral nervous system (PNS) and the central nervous system (CNS).

N-2 Supplement has been demonstrated in wide-ranging applications, such as the culture of neural stem cells, the culture of the SH-SY5Y cell line (subline of the neuroblastoma cell line SK-N-SH), and culture of the HT22 mouse hippocampal neuronal cell line. Additionally, N-2 Supplement adds support to cell culture media used to differentiate pluripotent stem cells (PSCs) to a variety of cell types, including neural and pancreatic-like cell types.

N-2 Supplement may be used with Neurobasal media supplemented with growth factors such as bFGF and EGF. When combined with Dulbecco's Modified Eagle Medium (D-MEM) and fetal bovine serum (FBS), N-2 Supplement also provides suitable supplementation for the culture of human and rodent astrocytes.

  • Each lot is cGMP manufactured
  • Each lot is tested for sterility, endotoxin, and performance
  • Contains insulin, iron-rich holo-transferrin, progesterone, putrescine, and selenite
  • Can be used as a substitute for Bottenstein's N-1 formulation
  • Provided as a 100X concentrate

Choose Gibco N-2 Supplement for a reliable, high-quality product trusted by researchers worldwide.

TRUSTED_SUSTAINABILITY

Specifica

Content And Storage Store in freezer (-5 to -20°C) and protect from light.

Cell Type Neural Cells
Classification Serum-free
Concentration 100 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
pH 6 to 8
Quantity 5 mL
Shipping Condition Dry Ice
Tested For Endotoxin, Performance, Sterility
Vedi altri risultati Mostra meno risultati
Identificativo prodotto
  • Mercuric acetate
Informazioni supplementari
  • MIXTURE LIST-Contains : Mercuric acetate

For Research Use Only. Not for use in diagnostic procedures.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato