missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Formyl-4-pyridinyl)-2,2-dimethylpropanamide, 97%, Thermo Scientific™

20.90 € - 46.70 €

Chemical Identifiers

CAS 86847-71-4
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD03086208
InChI Key ICMXCEJBHWHTBH-UHFFFAOYSA-N
Synonym n-3-formylpyridin-4-yl pivalamide, n-3-formyl-4-pyridinyl-2,2-dimethylpropanamide, n-3-formylpyridin-4-yl-2,2-dimethylpropanamide, n-3-formyl-pyridin-4-yl-2,2-dimethyl-propionamide, 4-2,2-dimethylpropanoyl amino pyridine-3-carboxaldehyde, 2,2-dimethyl-n-3-formylpyridin-4-yl propanamide, 4-2,2-dimethylpropanoyl amino nicotinaldehyde, propanamide,n-3-formyl-4-pyridinyl-2,2-dimethyl, 4-2,2-dimethylpropanoyl amino-3-formylpyridine, n-3-formylpyrdn-4-yl pvalamde
PubChem CID 2779664
IUPAC Name N-(3-formylpyridin-4-yl)-2,2-dimethylpropanamide
SMILES CC(C)(C)C(=O)NC1=C(C=NC=C1)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10703823
SDS View Documents
Thermo Scientific Maybridge
MO00095CB
250 mg Amber Glass Bottle
20.90 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10009291
SDS View Documents
Thermo Scientific Maybridge
MO00095DA
1 g Amber Glass Bottle
46.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(3-Formyl-4-pyridinyl)-2,2-dimethylpropanamide, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.