missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Carboxyphenyl)succinamic acid, 99%, Thermo Scientific Chemicals

Product Code. 11348496
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76475-62-2
Molecular Formula C11H11NO5
Molecular Weight (g/mol) 237.21
MDL Number MFCD00020530
InChI Key IOKXKKSBNJCKOY-UHFFFAOYSA-N
Synonym 4-3-carboxypropanamido benzoic acid, 4-3-carboxypropanoyl amino benzoic acid, n-4-carboxyphenyl succinamic acid, 4-3-carboxypropanoylamino benzoic acid, cbkinase1_000202, cbkinase1_012602, cambridge id 5122038, n-4-carboxyphenyl succinamidic acid, 4-2-carboxyethylcarboxamido benzoic acid
PubChem CID 765581
IUPAC Name 4-(3-carboxypropanoylamino)benzoic acid
SMILES OC(=O)CCC(=O)NC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point ∽290°C (decomposition)
Quantity 1 g
Beilstein 3355574
Formula Weight 237.21
Percent Purity 99%
Chemical Name or Material N-(4-Carboxyphenyl)succinamic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'