Learn More
N-(4-Chloro-2-butynyl)phthalimide, 97%, Thermo Scientific Chemicals
86.03 € - 322.61 €
Chemical Identifiers
CAS | 4819-69-6 |
---|---|
Molecular Formula | C12H8ClNO2 |
Molecular Weight (g/mol) | 233.651 |
MDL Number | MFCD01318120 |
InChI Key | TXNDRPKNOXQAAO-UHFFFAOYSA-N |
Synonym | n-4-chloro-2-butynyl phthalimide, 2-4-chlorobut-2-yn-1-yl isoindoline-1,3-dione, 2-4-chlorobut-2-yn-1-yl-1h-isoindole-1,3 2h-dione, 2-4-chlorobut-2-yn-1-yl isoindole-1,3-dione, acmc-20akbi, n-4-chlorobut-2-ynyl phthalimide, 2-4-chlorobut-2-ynyl isoindole-1,3-dione, 2-4-chloro-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-chloro-2-butyn-1-yl, 2-4-chlorobut-2-yn-1-yl-2,3-dihydro-1h-isoindole-1,3-dione |
PubChem CID | 237537 |
IUPAC Name | 2-(4-chlorobut-2-ynyl)isoindole-1,3-dione |
SMILES | C1=CC=C2C(=C1)C(=O)N(C2=O)CC#CCCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11312987
|
Thermo Scientific Alfa Aesar
L16313.03 |
1 g |
86.03 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11322987
|
Thermo Scientific Alfa Aesar
L16313.06 |
5 g |
322.61 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-(4-Chloro-2-butynyl)phthalimide is used to make other chemicals and as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-(4-Chloro-2-butynyl)phthalimide is used to make other chemicals and as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
Chemical Identifiers
4819-69-6 | |
233.651 | |
TXNDRPKNOXQAAO-UHFFFAOYSA-N | |
237537 | |
C1=CC=C2C(=C1)C(=O)N(C2=O)CC#CCCl |
C12H8ClNO2 | |
MFCD01318120 | |
n-4-chloro-2-butynyl phthalimide, 2-4-chlorobut-2-yn-1-yl isoindoline-1,3-dione, 2-4-chlorobut-2-yn-1-yl-1h-isoindole-1,3 2h-dione, 2-4-chlorobut-2-yn-1-yl isoindole-1,3-dione, acmc-20akbi, n-4-chlorobut-2-ynyl phthalimide, 2-4-chlorobut-2-ynyl isoindole-1,3-dione, 2-4-chloro-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-chloro-2-butyn-1-yl, 2-4-chlorobut-2-yn-1-yl-2,3-dihydro-1h-isoindole-1,3-dione | |
2-(4-chlorobut-2-ynyl)isoindole-1,3-dione |
Specifications
4819-69-6 | |
Nearly Odorless | |
MFCD01318120 | |
179626 | |
Insoluble in water. | |
C1=CC=C2C(=C1)C(=O)N(C2=O)CC#CCCl | |
233.651 | |
233.66 | |
N-(4-Chloro-2-butynyl)phthalimide |
115°C to 120°C | |
C12H8ClNO2 | |
UN3261 | |
n-4-chloro-2-butynyl phthalimide, 2-4-chlorobut-2-yn-1-yl isoindoline-1,3-dione, 2-4-chlorobut-2-yn-1-yl-1h-isoindole-1,3 2h-dione, 2-4-chlorobut-2-yn-1-yl isoindole-1,3-dione, acmc-20akbi, n-4-chlorobut-2-ynyl phthalimide, 2-4-chlorobut-2-ynyl isoindole-1,3-dione, 2-4-chloro-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-chloro-2-butyn-1-yl, 2-4-chlorobut-2-yn-1-yl-2,3-dihydro-1h-isoindole-1,3-dione | |
TXNDRPKNOXQAAO-UHFFFAOYSA-N | |
2-(4-chlorobut-2-ynyl)isoindole-1,3-dione | |
237537 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.