missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Cyanophenyl)thiourea, 98%, Thermo Scientific Chemicals

Product Code. 11398675
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3460-55-7
Molecular Formula C8H7N3S
Molecular Weight (g/mol) 177.225
MDL Number MFCD00041175
InChI Key LFMJROANUIRGBS-UHFFFAOYSA-N
Synonym 4-cyanophenyl thiourea, 1-4-cyanophenyl thiourea, 1-4-cyanophenyl-2-thiourea, n-4-cyanophenyl thiourea, 4-cyanophenyl-thiourea, acmc-1ajd4, thiourea,n-4-cyanophenyl, 4-aminothioxomethyl amino benzenecarbonitrile
PubChem CID 2757973
IUPAC Name (4-cyanophenyl)thiourea
SMILES C1=CC(=CC=C1C#N)NC(=S)N

Specifications

Melting Point ∽205°C (decomposition)
Quantity 5 g
UN Number UN3439
Beilstein 2718720
Formula Weight 177.23
Percent Purity 98%
Chemical Name or Material N-(4-Cyanophenyl)thiourea

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'