missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Hexynyl)phthalimide, 97%, Thermo Scientific Chemicals

Product Code. 15440368
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6097-08-1
MDL Number MFCD00671372

Specifications

Melting Point 68°C to 72°C
Molecular Formula C14H13NO2
Quantity 25 g
Synonym n-5-hexynyl phthalimide, 2-hex-5-yn-1-yl isoindoline-1,3-dione, 6-phthalimido-1-hexyne, 2-hex-5-yn-1-yl isoindole-1,3-dione, acmc-20akav, n-5-hexynylphthalimide, n-hex-5-ynylphthalimide, 6-phthalimidyl-1-hexyne
InChI Key WALVSJIWVWDFCU-UHFFFAOYSA-N
SMILES C#CCCCCN1C(=O)C2=CC=CC=C2C1=O
IUPAC Name 2-hex-5-ynylisoindole-1,3-dione
Molecular Weight (g/mol) 227.26
PubChem CID 3346053
Formula Weight 227.26
Percent Purity 97%
Chemical Name or Material N-(5-Hexynyl)phthalimide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'