Learn More
N-Acetyl-D-alanine, 98%, Thermo Scientific Chemicals
Description
N-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.
Solubility
Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.

Specifications
Specifications
Solubility Information | Slightly soluble in water. |
Quantity | 1 g |
Formula Weight | 131.13 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | N-Acetyl-D-alanine |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.