missing translation for 'onlineSavingsMsg'
Learn More
N-Acetyl-D-alanine, 98%, Thermo Scientific Chemicals Product Code.: 15480149

N-Acetyl-D-alanine, 98%, Thermo Scientific Chemicals

Product Code. 15480149
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15480149

Brand: Thermo Scientific Alfa Aesar H63450.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19436-52-3
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00063133
InChI Key KTHDTJVBEPMMGL-UHFFFAOYNA-N
Synonym n-acetyl-d-alanine, ac-d-ala-oh, r-2-acetamidopropanoic acid, acetyl-d-alanine, n-acetylalanine, 2r-2-acetamidopropanoic acid, r-2-acetylamino-propionic acid, alanine, n-acetyl, n-acetyl-d-ala, n-acetyl d-alanine
PubChem CID 132213
IUPAC Name (2R)-2-acetamidopropanoic acid
SMILES CC(NC(C)=O)C(O)=O

Specifications

Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 131.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Acetyl-D-alanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU23K026 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberH63450.06,H63450.03
Lot NumberT22K026 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog NumberH63450.06,H63450.03
Lot NumberM03J014 Certificate TypeCertificate of Analysis Date10/01/2023 Catalog NumberH63450.06,H63450.03
Lot NumberN20G031 Certificate TypeCertificate of Analysis Date13/07/2022 Catalog NumberH63450.06,H63450.03
Lot NumberN07G011 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH63450.06,H63450.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Acetyl-D-alanine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.