missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-phenylalanine, 95%, Thermo Scientific™

Product Code. 15450049
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10172-89-1
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00002664
InChI Key CBQJSKKFNMDLON-SNVBAGLBSA-N
Synonym n-acetyl-d-phenylalanine, acetyl-d-phenylalanine, ac-d-phe-oh, n-acetyl-3-phenyl-d-alanine, n-acetylphenylalanine, d, d-phenylalanine, n-acetyl, r-2-acetamido-3-phenylpropanoic acid, 2r-2-acetamido-3-phenylpropanoic acid, n-acetylphenylalanine #, n-ac-d-phe-oh
PubChem CID 101184
ChEBI CHEBI:28203
IUPAC Name (2R)-2-acetamido-3-phenylpropanoic acid
SMILES CC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 165°C to 169°C
Color White
Beilstein 2213852
Quantity 100 g
Formula Weight 207.23
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Acetyl-D-phenylalanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.