missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-alanine, 96%

Product Code. 11332637
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-69-8
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.131
MDL Number MFCD00063132
InChI Key KTHDTJVBEPMMGL-VKHMYHEASA-N
Synonym n-acetyl-l-alanine, ac-ala-oh, acetylalanine, acetyl-l-alanine, n-acetylalanine, 2-acetamidopropionic acid, l-n-acetylalanine, n-acetyl-s-alanine, l-alanine, n-acetyl, 2s-2-acetamidopropanoic acid
PubChem CID 88064
ChEBI CHEBI:40992
IUPAC Name (2S)-2-acetamidopropanoic acid
SMILES CC(C(=O)O)NC(=O)C

Specifications

Melting Point 122°C to 128°C
Beilstein 1722932
Quantity 1 g
Formula Weight 131.13
Optical Rotation −64° (c=2 in Water)
Percent Purity 96%
Chemical Name or Material N-Acetyl-L-alanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals N-Acetyl-L-alanine, 96% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.