missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Acetyl-L-leucine, 99%
35.78 € - 120.60 €
Chemical Identifiers
CAS | 1188-21-2 |
---|---|
Molecular Formula | C8H15NO3 |
Molecular Weight (g/mol) | 173.21 |
MDL Number | MFCD00065131 |
InChI Key | WXNXCEHXYPACJF-JLDDOWRYNA-N |
Synonym | n-acetyl-l-leucine, acetyl-l-leucine, ac-leu-oh, n-acetylleucine, s-2-acetamido-4-methylpentanoic acid, n-acetyl-leu, 2s-2-acetamido-4-methylpentanoic acid, n-acetyl-leucine, leucine, n-acetyl-, l, n-acetyl-l-leucin |
PubChem CID | 70912 |
ChEBI | CHEBI:17786 |
IUPAC Name | (2S)-2-acetamido-4-methylpentanoic acid |
SMILES | CC(C)C[C@H](NC(C)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10235961
|
Thermo Scientific Chemicals
347010050 |
5 g | Glass bottle |
35.78 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10377181
|
Thermo Scientific Chemicals
347010250 |
25 g | Glass bottle |
120.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1188-21-2 | |
173.21 | |
WXNXCEHXYPACJF-JLDDOWRYNA-N | |
70912 | |
(2S)-2-acetamido-4-methylpentanoic acid |
C8H15NO3 | |
MFCD00065131 | |
n-acetyl-l-leucine, acetyl-l-leucine, ac-leu-oh, n-acetylleucine, s-2-acetamido-4-methylpentanoic acid, n-acetyl-leu, 2s-2-acetamido-4-methylpentanoic acid, n-acetyl-leucine, leucine, n-acetyl-, l, n-acetyl-l-leucin | |
CHEBI:17786 | |
CC(C)C[C@H](NC(C)=O)C(O)=O |
Specifications
1188-21-2 | |
100.0 | |
White | |
99% | |
(CH3)2CHCH2CH(NHCOCH3)COOH | |
− 24.50 | |
(5% in ethanol) Clear colorless | |
CC(C)C[C@H](NC(C)=O)C(O)=O | |
173.21 | |
CHEBI:17786 | |
99% | |
N-Acetyl-L-leucine, 99% |
98.5 | |
190.0°C | |
Authentic | |
C8H15NO3 | |
MFCD00065131 | |
n-acetyl-l-leucine, acetyl-l-leucine, ac-leu-oh, n-acetylleucine, s-2-acetamido-4-methylpentanoic acid, n-acetyl-leu, 2s-2-acetamido-4-methylpentanoic acid, n-acetyl-leucine, leucine, n-acetyl-, l, n-acetyl-l-leucin | |
WXNXCEHXYPACJF-JLDDOWRYNA-N | |
(2S)-2-acetamido-4-methylpentanoic acid | |
70912 | |
173.21 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 214-706-3
missing translation for 'tsca' : TSCA
Product Identifier
- N-Acetyl-L-leucine
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title