missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-tyrosine, 99%

Product Code. 11463547
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Solubility
Soluble in water (25 mg/ml), and ethanol.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-55-3
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00037190
InChI Key CAHKINHBCWCHCF-UEQNJFAPNA-N
Synonym n-acetyl-l-tyrosine, ac-tyr-oh, acetyl-l-tyrosine, n-acetyl-tyrosine, n-acetyltyrosine, acetyl tyrosine, l-n-acetyltyrosine, tyrosine, n-acetyl, l-tyrosine, n-acetyl, unii-da8g610zo5
PubChem CID 68310
ChEBI CHEBI:21563
IUPAC Name (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point 150°C to 151°C
Beilstein 2697172
Solubility Information Soluble in water (25mg/ml),and ethanol.
Quantity 250 g
Formula Weight 223.23
Optical Rotation +58° (c=5 in Ethanol)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-tyrosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals N-Acetyl-L-tyrosine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.