missing translation for 'onlineSavingsMsg'
Learn More

N-Acryloxysuccinimide, 99%, Thermo Scientific Chemicals

113.57 € - 1626.09 €

Chemical Identifiers

CAS 38862-24-7
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00078261
InChI Key YXMISKNUHHOXFT-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl acrylate, n-acryloxysuccinimide, n-succinimidyl acrylate, acrylic acid n-hydroxysuccinimide ester, 1-acryloyloxy-2,5-pyrrolidinedione, n-acryloyloxysuccinimide, 2,5-pyrrolidinedione, 1-1-oxo-2-propenyl oxy, 2,5-dioxopyrrolidin-1-yl prop-2-enoate, poly n-acryloxysuccinimide
PubChem CID 181508
IUPAC Name (2,5-dioxopyrrolidin-1-yl) prop-2-enoate
SMILES C=CC(=O)ON1C(=O)CCC1=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10085702
SDS View Documents
Thermo Scientific Acros
400300010
1 g Glass bottle
113.57 €
1g
Please to purchase this item. Need a web account? Register with us today!
10527172
SDS View Documents
Thermo Scientific Acros
400300050
5 g Glass bottle
372.86 €
5g
Please to purchase this item. Need a web account? Register with us today!
11976911
SDS View Documents
Thermo Scientific Acros
400300250
25 g Glass bottle
1626.09 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Chemical Identifiers

38862-24-7
169.14
YXMISKNUHHOXFT-UHFFFAOYSA-N
181508
C=CC(=O)ON1C(=O)CCC1=O
C7H7NO4
MFCD00078261
2,5-dioxopyrrolidin-1-yl acrylate, n-acryloxysuccinimide, n-succinimidyl acrylate, acrylic acid n-hydroxysuccinimide ester, 1-acryloyloxy-2,5-pyrrolidinedione, n-acryloyloxysuccinimide, 2,5-pyrrolidinedione, 1-1-oxo-2-propenyl oxy, 2,5-dioxopyrrolidin-1-yl prop-2-enoate, poly n-acryloxysuccinimide
(2,5-dioxopyrrolidin-1-yl) prop-2-enoate

Specifications

38862-24-7
Yellow
99%
MFCD00078261
Solubility in water: soluble. Other solubilities: soluble in diethyl ether,hexane,ethyl acetate
C=CC(=O)ON1C(=O)CCC1=O
169.14
169.14
Crystals and/or Chunks
69.0°C
Authentic
C7H7NO4
2,5-dioxopyrrolidin-1-yl acrylate, n-acryloxysuccinimide, n-succinimidyl acrylate, acrylic acid n-hydroxysuccinimide ester, 1-acryloyloxy-2,5-pyrrolidinedione, n-acryloyloxysuccinimide, 2,5-pyrrolidinedione, 1-1-oxo-2-propenyl oxy, 2,5-dioxopyrrolidin-1-yl prop-2-enoate, poly n-acryloxysuccinimide
YXMISKNUHHOXFT-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) prop-2-enoate
181508
99%
N-Acryloxysuccinimide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Product Identifier
  • N-Acryloxysuccinimide

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.