Learn More
N-Allylmethylamine, 96%, Thermo Scientific Chemicals
36.58 € - 119.60 €
Chemical Identifiers
CAS | 627-37-2 |
---|---|
Molecular Formula | C4H9N |
Molecular Weight (g/mol) | 71.123 |
MDL Number | MFCD00008641 |
InChI Key | IOXXVNYDGIXMIP-UHFFFAOYSA-N |
Synonym | n-allylmethylamine, n-methylallylamine, 2-propen-1-amine, n-methyl, allylmethylamine, methylallylamine, n-methyl-2-propen-1-amine, n-allyl-n-methylamine, methyl prop-2-en-1-yl amine, allylamine, n-methyl-7ci,8ci, 2-propen-1-amine, n-methyl-9ci |
PubChem CID | 69391 |
IUPAC Name | N-methylprop-2-en-1-amine |
SMILES | CNCC=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11421850
|
Thermo Scientific Alfa Aesar
H27551.03 |
1 g |
36.58 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431850
|
Thermo Scientific Alfa Aesar
H27551.06 |
5 g |
119.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Allylmethylamine is used in the production of N-allyl-alfa,alfa-dichloro-N-methylacetamide. It is used as a functional monomer to coordinate with Cd(II)/Zn(II) ions to get the poly methyl methacrylate -butyl methacrylate - N-allylmethylamine (PMBA) films, which is useful as building blocks to fabricate a photo-catalytic cell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Allylmethylamine is used in the production of N-allyl-alfa,alfa-dichloro-N-methylacetamide. It is used as a functional monomer to coordinate with Cd(II)/Zn(II) ions to get the poly methyl methacrylate -butyl methacrylate - N-allylmethylamine (PMBA) films, which is useful as building blocks to fabricate a photo-catalytic cell.
Solubility
Miscible with water.
Notes
Store in a cool place. Incompatible with oxidizing agents, strong acids and bases.
Chemical Identifiers
627-37-2 | |
71.123 | |
IOXXVNYDGIXMIP-UHFFFAOYSA-N | |
69391 | |
CNCC=C |
C4H9N | |
MFCD00008641 | |
n-allylmethylamine, n-methylallylamine, 2-propen-1-amine, n-methyl, allylmethylamine, methylallylamine, n-methyl-2-propen-1-amine, n-allyl-n-methylamine, methyl prop-2-en-1-yl amine, allylamine, n-methyl-7ci,8ci, 2-propen-1-amine, n-methyl-9ci | |
N-methylprop-2-en-1-amine |
Specifications
627-37-2 | |
61°C to 62°C | |
C4H9N | |
MFCD00008641 | |
n-allylmethylamine, n-methylallylamine, 2-propen-1-amine, n-methyl, allylmethylamine, methylallylamine, n-methyl-2-propen-1-amine, n-allyl-n-methylamine, methyl prop-2-en-1-yl amine, allylamine, n-methyl-7ci,8ci, 2-propen-1-amine, n-methyl-9ci | |
IOXXVNYDGIXMIP-UHFFFAOYSA-N | |
N-methylprop-2-en-1-amine | |
69391 | |
96% |
0.748 | |
−8°C (−18°F) | |
1.412 | |
UN2733 | |
Miscible with water. | |
CNCC=C | |
71.123 | |
71.12 | |
N-Allylmethylamine |
Safety and Handling
GHS H Statement
H225-H301-H311-H314-H318-H332
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H301+H311+H331-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 210-996-0
missing translation for 'rtecsNumber' : UC6632500
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.