missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals N(alpha)-Boc-L-asparagine, 98+% Product Code.: 11428646

Thermo Scientific Chemicals N(alpha)-Boc-L-asparagine, 98+%

Product Code. 11428646
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11428646

Brand: Thermo Scientific Chemicals A16019.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7536-55-2
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00038152
InChI Key FYYSQDHBALBGHX-YFKPBYRVSA-N
Synonym boc-asn-oh, boc-l-asparagine, boc-asn, n-tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonylasparagine, nalpha-tert-butoxycarbonyl-l-asparagine, boc-asparagine, tert-butoxycarbonyl-l-asparagine, n-tert-butoxycarbonyl asparagine, tert-butyloxycarbonyl-l-asparagine
PubChem CID 82035
ChEBI CHEBI:3146
IUPAC Name (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O

Specifications

Melting Point ∼170°C (decomposition)
Beilstein 1977963
Quantity 5 g
Formula Weight 232.24
Optical Rotation −7.3° (c=2 in DMF)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Boc-L-asparagine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10213276 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16019.14,A16019.06
Lot Number10234445 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16019.14,A16019.06
Lot Number10233021 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16019.14,A16019.06
Lot Number10178619 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16019.14,A16019.06
Lot NumberB28Y014 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16019.14,A16019.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals N(alpha)-Boc-L-asparagine, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.