missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ N(alpha)-Ethoxycarbonyl-L-asparagine, 97%

Product Code. 11377346
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 16639-91-1
Molecular Formula C7H12N2O5
Molecular Weight (g/mol) 204.182
MDL Number MFCD00082761
InChI Key YORYXPTWPIKBDE-BYPYZUCNSA-N
Synonym eoc-l-asparagine, l-asparagine, n2-ethoxycarbonyl, n alpha-ethoxycarbonyl-l-asparagine, s-4-amino-2-ethoxycarbonyl amino-4-oxobutanoic acid, n-carbethoxy-l-asparagine, n∼2∼-ethoxycarbonyl-l-asparagine, 2s-3-carbamoyl-2-ethoxycarbonyl amino propanoic acid, n a ethoxycarbonyl-l-asparagine, l-asparagine,n2-ethoxycarbonyl, n alpha-carbethoxy-l-asparagine
PubChem CID 7021476
IUPAC Name (2S)-4-amino-2-(ethoxycarbonylamino)-4-oxobutanoic acid
SMILES CCOC(=O)NC(CC(=O)N)C(=O)O

Specifications

Melting Point ∽166°C (decomposition)
Beilstein 1712104
Quantity 1 g
Formula Weight 204.18
Percent Purity 97%
Chemical Name or Material N(alpha)-Ethoxycarbonyl-L-asparagine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'