missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(alpha)-Fmoc-L-asparagine, 98%

Product Code. 11470087
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-16-7
Molecular Formula C19H18N2O5
Molecular Weight (g/mol) 354.362
MDL Number MFCD00037132
InChI Key YUGBZNJSGOBFOV-INIZCTEOSA-N
Synonym fmoc-asn-oh, fmoc-l-asn-oh, fmoc-l-asparagine, nalpha-fmoc-l-asparagine, n-fmoc-l-asparagine, fmoc-asn, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-asparagine, n-a-fmoc-l-asparagine, 9-fluorenylmethoxycarbonyl-l-asparagine, 2s-3-carbamoyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 2724774
IUPAC Name (2S)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxobutanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)N)C(=O)O

Specifications

Melting Point ∼188°C (decomposition)
Beilstein 4335103
Quantity 5 g
Formula Weight 354.36
Optical Rotation −12±1° (c=1 in DMF)
Percent Purity 98%
Chemical Name or Material N(alpha)-Fmoc-L-asparagine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'