missing translation for 'onlineSavingsMsg'
Learn More
N-Benzoyl-2'-deoxycytidine, 98+%, Thermo Scientific Chemicals Product Code.: 15428927

N-Benzoyl-2'-deoxycytidine, 98+%, Thermo Scientific Chemicals

Product Code. 15428927
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15428927

Brand: Thermo Scientific Alfa Aesar H52298.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine, N-Protected 2?-Deoxyribonucleosides(useful for oligonucleotide synthesis), 3?-, 5?-and 3?, 5?-di-O-crotonyl 2?-deoxynucleoside derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4836-13-9
Molecular Formula C16H17N3O5
Molecular Weight (g/mol) 331.33
MDL Number MFCD00010115
InChI Key MPSJHJFNKMUKCN-OUCADQQQSA-N
Synonym n4-benzoyl-2'-deoxycytidine, n-benzoyl-2'-deoxy-cytidine, n-benzoyl-2'-deoxycytidine, cytidine, n-benzoyl-2'-deoxy, cytidine,n-benzoyl-2'-deoxy, n-benzoyl-2-deoxy-cytidine, dc-bz, ksc590o7t, n-benzoyl-2/'-deoxycytidine
PubChem CID 9797617
IUPAC Name N-[1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O

Specifications

Packaging Glass bottle
Quantity 5 g
Solubility Information Soluble in methanol (50mg/ml).
Formula Weight 331.33
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material N-Benzoyl-2′-deoxycytidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Benzoyl-2'-deoxycytidine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.