missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride, 98%

Product Code. 10732081
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 911-77-3
Molecular Formula C19H23ClN6O4
Molecular Weight (g/mol) 434.881
MDL Number MFCD00012846
InChI Key DEOKFPFLXFNAON-UHFFFAOYSA-N
Synonym bapna, n-5-guanidino-1-4-nitrophenyl amino-1-oxopentan-2-yl benzamide hydrochloride, n-benzoyl-dl-arginine-4-nitroanilide hydrochloride, n-alpha-benzoyl-dl-arginine-4-nitroanilide hydrochloride, nalpha-benzoyl-dl-arginine 4-nitroanilide hydrochloride, bani, 5-carbamimidamido-n-4-nitrophenyl-2-phenylformamido pentanamide hydrochloride, l-bapna, dl-bapa, bz-dl-arg-pna hcl
PubChem CID 2724371
IUPAC Name N-[5-(diaminomethylideneamino)-1-(4-nitroanilino)-1-oxopentan-2-yl]benzamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)NC2=CC=C(C=C2)[N+](=O)[O-].Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass Bottle
Formula Weight 434.89
Percent Purity ≥97.5% (HPLC)
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
Show More Show Less
Product Identifier
  • N-a-Benzoyl-DL-arginine-4-nitroanilide hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'