missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-4-(2-hydroxyethyl)piperidine, 97%, Thermo Scientific™

Product Code. 10753331
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 76876-70-5
CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C14H21NO
Refractive Index 1.5370 to 1.5400
MDL Number MFCD00800492
Quantity 1 g
Synonym 2-1-benzylpiperidin-4-yl ethanol, n-benzyl-4-2-hydroxyethyl piperidine, 1-benzyl-4-2-hydroxyethyl piperdine, 1-benzyl-4-2-hydroxyethyl piperidine, 2-1-benzyl-4-piperidyl ethan-1-ol, 1-benzylpiperidine-4-ethanol, 1-phenylmethyl-4-piperidineethanol, 4-piperidineethanol,1-phenylmethyl, 2-1-benzylpiperidin-4-yl ethan-1-ol, n-benzyl-4-2-hydroxyethyl-piperidine
InChI Key FBFPTWSKNJHCGT-UHFFFAOYSA-N
SMILES C1CN(CCC1CCO)CC2=CC=CC=C2
IUPAC Name 2-(1-benzylpiperidin-4-yl)ethanol
Molecular Weight (g/mol) 219.33
PubChem CID 2725029
Formula Weight 219.33
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material N-Benzyl-4-(2-hydroxyethyl)piperidine, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'