missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-4-chlorobenzamide, 97%, Thermo Scientific™

Código de producto. 15401808
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7461-34-9
Molecular Formula C14H12ClNO
Molecular Weight (g/mol) 245.71
MDL Number MFCD00018682
InChI Key LSMWDKIFKGLNSW-UHFFFAOYSA-N
Synonym n-benzyl-4-chloro-benzamide, 4-chloro-n-benzylbenzamide, n∼1∼-benzyl-4-chlorobenzamide, 4-chloro-n-phenylmethyl benzamide, 4-chlorophenyl-n-benzylcarboxamide, 4-chloranyl-n-phenylmethyl benzamide
PubChem CID 346668
IUPAC Name N-benzyl-4-chlorobenzamide
SMILES ClC1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1

Specifications

Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 245.71
Percent Purity 97%
Chemical Name or Material N-Benzyl-4-chlorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'