missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-N-phenylhydrazine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11483607
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5705-15-7
Molecular Formula C13H15ClN2
Molecular Weight (g/mol) 234.73
MDL Number MFCD00050690
InChI Key JTYLHYOCBGPMNO-UHFFFAOYSA-N
Synonym 1-benzyl-1-phenylhydrazine hydrochloride, n-benzyl-n-phenylhydrazine hydrochloride, usaf ek-1666, hydrazine, 1-phenyl-1-phenylmethyl-, monohydrochloride, alpha-benzyl-alpha-phenylhydrazine hydrochloride, n-benzyl-n-phenylhydrazinium 1+ chloride, hydrazine, 1-benzyl-1-phenyl-, hydrochloride, 1-benzyl-1-phenylhydrazine hydrochloride van, hydrazine, 1-phenyl-1-phenylmethyl-, hydrochloride 1:1, acmc-209lvq
PubChem CID 79785
IUPAC Name 1-benzyl-1-phenylhydrazine;hydrochloride
SMILES [H+].[Cl-].NN(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 178°C to 181°C
Quantity 25 g
Beilstein 3707186
Formula Weight 234.73
Percent Purity ≥98%
Chemical Name or Material N-Benzyl-N-phenylhydrazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'