missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyl-tert-butylamine, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Specifications
Specifications
Melting Point | -30°C to -25°C |
Density | 0.899 |
Boiling Point | 218°C to 220°C |
Flash Point | 88°C (190°F) |
Linear Formula | C6H5CH2NHC(CH3)3 |
Refractive Index | 1.497 |
Quantity | 25 g |
UN Number | UN2735 |
Beilstein | 507464 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-Benzyl-tert-butylamine, 96%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction