missing translation for 'onlineSavingsMsg'
Learn More
N-Benzylglycine hydrochloride, 98+%, Thermo Scientific Chemicals Product Code.: 11398206

N-Benzylglycine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11398206
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11398206

Brand: Thermo Scientific Alfa Aesar L15200.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7689-50-1
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00156934
InChI Key BUZJPENZWLUHJD-UHFFFAOYSA-N
Synonym n-benzylglycine hydrochloride, benzylaminoacetic acid hydrochloride, 2-benzylamino acetic acid hydrochloride, bzl-gly-oh.hcl, n-benzylglycine hcl, bz-gly-oh.hcl, n-benzyl glycine hydrochloride, bzl-gly-oh inverted exclamation mark currencyhcl, benzylamino acetic acid hydrochloride, glycine, n-phenylmethyl-, hydrochloride
PubChem CID 2756624
IUPAC Name 2-(benzylamino)acetic acid;hydrochloride
SMILES C1=CC=C(C=C1)CNCC(=O)O.Cl

Specifications

Melting Point ∽220°C (decomposition)
Beilstein 3915652
Quantity 100 g
Formula Weight 201.65
Percent Purity ≥98%
Chemical Name or Material N-Benzylglycine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10126526 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberL15200.06,L15200.14,L15200.22
Lot Number10106200 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberL15200.06,L15200.14,L15200.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Benzylglycine hydrochloride, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.