missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-alaninol, 98%, Thermo Scientific Chemicals

Code produit. 11470467
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 61425-27-2
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00672531
InChI Key AFPHMHSLDRPUSM-SECBINFHSA-N
Synonym r-benzyl 1-hydroxypropan-2-yl carbamate, cbz-d-alaninol, z-d-ala-ol, n-z-d-alaninol, r-2-cbz-amino-1-propanol, n-benzyloxycarbonyl-d-alaninol, benzyl n-2r-1-hydroxypropan-2-yl carbamate, benzyl 1r-2-hydroxy-1-methylethylcarbamate, n-1r-2-hydroxy-isopropyl phenylmethoxy carboxamide
PubChem CID 6951254
IUPAC Name benzyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
SMILES C[C@H](CO)NC(=O)OCC1=CC=CC=C1

Spécification

Melting Point 82°C to 84°C
Quantity 25 g
Formula Weight 209.25
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-D-alaninol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'