missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-threonine, 98+%, Thermo Scientific™

15.00 € - 24.80 €

Chemical Identifiers

CAS 80384-27-6
Molecular Formula C12H15NO5
Molecular Weight (g/mol) 253.25
MDL Number MFCD00037808
InChI Key IPJUIRDNBFZGQN-WCBMZHEXSA-N
Synonym z-d-thr-oh, z-d-threonine, n-carbobenzoxy-d-threonine, 2r,3s-2-benzyloxy carbonyl amino-3-hydroxybutanoic acid, n-benzyloxycarbonyl-d-threonine, n-cbz-d-threonine, d-threonine, n-phenylmethoxy carbonyl, 2r,3s-3-hydroxy-2-phenylmethoxycarbonylamino butanoic acid, cbz-d-thr-oh
PubChem CID 853484
IUPAC Name (2R,3S)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES C[C@H](O)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11322586
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L08561.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11312586
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L08561.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

80384-27-6
253.25
IPJUIRDNBFZGQN-WCBMZHEXSA-N
853484
C[C@H](O)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
C12H15NO5
MFCD00037808
z-d-thr-oh, z-d-threonine, n-carbobenzoxy-d-threonine, 2r,3s-2-benzyloxy carbonyl amino-3-hydroxybutanoic acid, n-benzyloxycarbonyl-d-threonine, n-cbz-d-threonine, d-threonine, n-phenylmethoxy carbonyl, 2r,3s-3-hydroxy-2-phenylmethoxycarbonylamino butanoic acid, cbz-d-thr-oh
(2R,3S)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoic acid

Specifications

80384-27-6
C12H15NO5
4695777
IPJUIRDNBFZGQN-WCBMZHEXSA-N
C[C@H](O)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
253.25
253.26
N-Benzyloxycarbonyl-D-threonine
100°C to 105°C
MFCD00037808
z-d-thr-oh, z-d-threonine, n-carbobenzoxy-d-threonine, 2r,3s-2-benzyloxy carbonyl amino-3-hydroxybutanoic acid, n-benzyloxycarbonyl-d-threonine, n-cbz-d-threonine, d-threonine, n-phenylmethoxy carbonyl, 2r,3s-3-hydroxy-2-phenylmethoxycarbonylamino butanoic acid, cbz-d-thr-oh
+5.5° (c=2 in Acetic Acid)
(2R,3S)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoic acid
853484
≥98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications