missing translation for 'onlineSavingsMsg'
Learn More
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals Product Code.: 15408066

N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals

Product Code. 15408066
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15408066

Brand: Thermo Scientific Alfa Aesar H62975.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.

Solubility
Soluble in methanol, dimethyl sulfoxide. Slightly soluble in water.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3705-42-8
Molecular Formula C20H21NO6
Molecular Weight (g/mol) 371.389
MDL Number MFCD00077013
InChI Key VWHKODOUMSMUAF-KRWDZBQOSA-N
Synonym z-glu-obzl, cbz-glu-obzl, cbz-l-glutamic acid 1-benzyl ester, z-l-glu-obzl, n-cbz-l-glutamic acid benzyl ester, n-carbobenzyloxy-l-glutamic acid o-benzyl ester, s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 4s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, n-benzyloxycarbonyl-l-glutamic acid, a-benzyl ester, z-l-glutamic acid alpha-benzyl ester
PubChem CID 6994031
IUPAC Name (4S)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 98°C to 102°C
Quantity 5 g
Solubility Information Soluble in methanol,dimethyl sulfoxide. Slightly soluble in water.
Formula Weight 371.38
Percent Purity 95%
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM28I074 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberH62975.14,H62975.06
Lot Number81500334 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62975.14,H62975.06
Lot NumberQ18F045 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62975.14,H62975.06
Lot NumberN09D007 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62975.14,H62975.06
Lot NumberX24F023 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62975.14,H62975.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.