missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-serine benzyl ester, 99%

Product Code. 11495257
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. Rückgaberichtlinie anzeigen

Artikelnummer. 11495257

Marke: Thermo Scientific Alfa Aesar B22647.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 21209-51-8 | C18H19NO5 | 329.352 g/mol

N-Benzyloxycarbonyl-L-serine benzyl ester is used as a building block in peptide synthesis and as a biochemical reagent. It is also used to synthesize the corresponding N-benzyloxycarbonyl-O-tert-butyl-(lor dl)-serine benzyl ester. Further, it serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-serine benzyl ester is used as a building block in peptide synthesis and as a biochemical reagent. It is also used to synthesize the corresponding N-benzyloxycarbonyl-O-tert-butyl-(lor dl)-serine benzyl ester. Further, it serves as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 21209-51-8
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.352
MDL Number MFCD00037824
InChI Key MHHDPGHZHFJLBZ-INIZCTEOSA-N
Synonym z-ser-obzl, cbz-ser-obzl, n-cbz-l-serine benzyl ester, n-z-l-serine benzyl ester, s-benzyl 2-benzyloxy carbonyl amino-3-hydroxypropanoate, n-carbobenzoxy-l-serine benzyl ester, benzyl n-benzyloxy carbonyl-l-serinate, l-serine,n-phenylmethoxy carbonyl-, phenylmethyl ester, serine, n-phenylmethoxy carbonyl-, phenylmethyl ester, z-l-serine benzyl ester
PubChem CID 6992970
IUPAC Name benzyl (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoate
SMILES C1=CC=C(C=C1)COC(=O)C(CO)NC(=O)OCC2=CC=CC=C2

Spezifikation

Melting Point 82°C to 84°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 329.32
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-serine benzyl ester

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado