missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyltropinone, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 215.29 |
Percent Purity | 97% |
Chemical Name or Material | 8-Benzyl-8-azabicyclo[3.2.1]octan-3-one |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction