missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-propargyl-L-glycine, 95%, Thermo Scientific™

Product Code. 15458647
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-2-propargyl-L-glycine is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63039-48-5
Molecular Formula C10H15NO4
Molecular Weight (g/mol) 213.233
MDL Number MFCD01320855
InChI Key AMKHAJIFPHJYMH-ZETCQYMHSA-N
Synonym boc-l-propargylglycine, s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-pra-oh, boc-propargyl-gly-oh, n-boc-2-propargyl-l-glycine, 2s-2-tert-butoxycarbonylamino pent-4-ynoic acid, 2s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, 4-pentynoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s, boc-propargylglycine, boc-l-propargyl glycine
PubChem CID 2734488
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid
SMILES CC(C)(C)OC(=O)NC(CC#C)C(=O)O

Specifications

Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 213.23
Percent Purity 95%
Chemical Name or Material N-Boc-2-propargyl-L-glycine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'