missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-aniline, 97%, Thermo Scientific Chemicals

Product Code. 15411418
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-3-3422
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD00043723
InChI Key KZZHPWMVEVZEFG-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC1=CC=CC=C1

Specifications

Melting Point 133°C to 137°C
Color Brown to White
Linear Formula C6H5NHCO2C(CH3)3
Quantity 5 g
Synonym n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester
IUPAC Name tert-butyl N-phenylcarbamate
PubChem CID 137930
Formula Weight 193.25
Percent Purity 0.97
Physical Form Solid
Chemical Name or Material N-Boc-aniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'