missing translation for 'onlineSavingsMsg'
Learn More
N-Boc-D-aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals Product Code.: 15418758

N-Boc-D-aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals

Product Code. 15418758
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15418758

Brand: Thermo Scientific Alfa Aesar H62210.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92828-64-3
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.35
MDL Number MFCD00038264
InChI Key LDRWTKQWSXGSTM-UHFFFAOYNA-N
Synonym boc-d-asp-obzl, r-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 3r-4-benzyloxy-3-tert-butoxy carbonyl amino-4-oxobutanoic acid, 3r-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, d-aspartic acid,n-1,1-dimethylethoxy carbonyl-, 1-phenylmethyl ester, boc-d-aspartic acid a-benzyl ester, boc-d-aspartic acid-1-benzyl ester, boc-d-aspartic acid, a-benzyl ester, boc-d-aspartic acid benzylester, n-tert-butoxycarbonyl-d-aspartic acid 1-benzyl ester
PubChem CID 11759109
SMILES CC(C)(C)OC(=O)NC(CC(O)=O)C(=O)OCC1=CC=CC=C1

Specifications

Color White
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
Quantity 5 g
Formula Weight 323.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-D-aspartic acid 1-benzyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-D-aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.