missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-threonine, 95%, Thermo Scientific Chemicals

Product Code. 15450449
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is known to inhibit growth and cell wall synthesis of Mycobacterium smegmatis. Also used as a synthetic intermediate for the production of chiral antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is known to inhibit growth and cell wall synthesis of Mycobacterium smegmatis. Also used as a synthetic intermediate for the production of chiral antibiotics.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55674-67-4
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.24
MDL Number MFCD00037807
InChI Key LLHOYOCAAURYRL-NTSWFWBYSA-N
Synonym boc-d-thr-oh, boc-d-threonine, n-tert-butoxycarbonyl-d-threonine, n-boc-d-threonine, 2r,3s-2-tert-butoxycarbonyl amino-3-hydroxybutanoic acid, n-alpha-tert-butoxycarbonyl-d-threonine, d-threonine, n-1,1-dimethylethoxy carbonyl, pubchem14945, boc-d-thr
PubChem CID 6992541
IUPAC Name (2R,3S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES C[C@H](O)[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Beilstein 2051734
Solubility Information Slightly soluble in water.
Quantity 25 g
Formula Weight 219.23
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-D-threonine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-D-threonine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.