missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-D-Valinol, 98%, Thermo Scientific™

Product Code. 10011224
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Infrared Spectrum Authentic
Color Colorless
CAS 106391-87-1
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Specific Rotation +22.5° to +23.5° (20°C, 589nm) (c=1 in CHCl3)
Boiling Point 218.0°C
Melting Point 57.0°C
Quantity 1 g
Physical Form Liquid
Assay Percent Range 98%
Molecular Formula C10H21NO3
MDL Number MFCD00235960
Synonym boc-d-valinol, n-boc-d-valinol, r-tert-butyl 1-hydroxy-3-methylbutan-2-yl carbamate, tert-butyl n-2r-1-hydroxy-3-methylbutan-2-yl carbamate, boc-d-val-ol, tert-butyl n-1r-1-hydroxymethyl-2-methyl-propyl carbamate, n-tert-butoxycarbonyl-d-valinol, n-t-boc-d-valinol, ksc491q4r
InChI Key OOQRRYDVICNJGC-QMMMGPOBSA-N
SMILES CC(C)[C@H](CO)NC(=O)OC(C)(C)C
IUPAC Name tert-butyl N-[(2R)-1-hydroxy-3-methylbutan-2-yl]carbamate
Molecular Weight (g/mol) 203.28
PubChem CID 11241137
Formula Weight 203.28
Percent Purity 98%
Chemical Name or Material N-BOC-D-Valinol
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-D-Valinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.