missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-alaninamide, 96%, Thermo Scientific Chemicals

Product Code. 15419118
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Boc-L-alaninamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-alaninamide is used as pharmaceutical intermediate.

Solubility
Soluble in water or 1% acetic acid.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85642-13-3
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD03701447
InChI Key GZKKSKKIABUUCX-UHFFFAOYNA-N
Synonym boc-ala-nh2, n-t-butyloxycarbonyl-l-alanine amide, s-tert-butyl 1-amino-1-oxopropan-2-yl carbamate, tert-butyl n-1s-1-carbamoylethyl carbamate, boc-beta-ala-nh2, boc-l-ala-nh2, boc-l-alanine-nh2, boc-l-alanine amide, s-boc-alaninamide, ambotzbaa5310
PubChem CID 7015295
SMILES CC(NC(=O)OC(C)(C)C)C(N)=O

Specifications

Quantity 250 mg
Solubility Information Soluble in water or 1% acetic acid.
IUPAC Name tert-butyl N-(1-carbamoylethyl)carbamate
Formula Weight 188.22
Percent Purity 96%
Physical Form Solid
Chemical Name or Material N-Boc-L-alaninamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.