missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-serine benzyl ester, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
It is a pharmaceutical intermediate. It is involved in Mitsunobu Reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a pharmaceutical intermediate. It is involved in Mitsunobu Reactions.
Solubility
Slightly soluble in water.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
It is a pharmaceutical intermediate. It is involved in Mitsunobu Reactions.
Solubility
Slightly soluble in water.
Notes
Store at room temperature. Keep away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 70°C |
Quantity | 5 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 295.33 |
Percent Purity | 95% |
Chemical Name or Material | N-Boc-L-serine benzyl ester |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-Boc-L-serine benzyl ester, 95%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction